accessibility menu, dialog, popup

UserName

CAS RN 5660-91-3

O

CAS RN 5660-91-3

IUPAC Name: 1,3-diphenyl-2H-cyclopenta[l]phenanthren-2-one
Synonyms: 1,3-diphenylcyclopenta[l]phenanthren-2-one
Molecular Weight (g/mol): 382.46
Molecular Formula: C29H18O
InChi Key: MNSDGJFEKUKHGO-UHFFFAOYSA-N
SMILES: O=C1C(=C2C(=C1C1=CC=CC=C1)C1=CC=CC=C1C1=CC=CC=C21)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results