accessibility menu, dialog, popup

UserName

CH3ONH2

CAS RN 56705-83-0

IUPAC Name: 4-(2-methylphenoxy)aniline
Synonyms: 4-(2-methylphenoxy)aniline
Molecular Weight (g/mol): 199.25
Molecular Formula: C13H13NO
InChi Key: JPCCVWJJMUIBJR-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1OC1=CC=C(N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results