accessibility menu, dialog, popup

UserName

NH2OCl

CAS RN 56705-85-2

IUPAC Name: 4-(2-chlorophenoxy)aniline
Synonyms: 4-(2-chlorophenoxy)aniline
Molecular Weight (g/mol): 219.67
Molecular Formula: C12H10ClNO
InChi Key: GKVDUWJVFNUYAR-UHFFFAOYSA-N
SMILES: NC1=CC=C(OC2=CC=CC=C2Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results