CAS RN 56806-74-7
CAS RN 56806-74-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Diphenyl p-Hydroxyphenyl Phosphate, TRC
CAS: 56806-74-7 Molecular Formula: C18H15O5P Molecular Weight (g/mol): 342.28 Synonym: Hydroquinonyldiphenyl Phosphate,Diphenyl p-Hydroxyphenylphosphoric Acid Ester; SMILES: Oc1ccc(OP(=O)(Oc2ccccc2)Oc3ccccc3)cc1