CAS RN 56843-79-9
CAS RN 56843-79-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Chloro-2-methylthieno[2,3-d]pyrimidine, TRC
CAS: 56843-79-9 Molecular Formula: C7H5ClN2S Molecular Weight (g/mol): 184.65 IUPAC Name: 4-chloro-2-methylthieno[2,3-d]pyrimidine SMILES: Cc1nc(Cl)c2ccsc2n1