accessibility menu, dialog, popup

UserName

BOOCH3CH3CH3CH3H3CCH3

CAS RN 569343-09-5

IUPAC Name: 2-(9,9-dimethyl-9H-fluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(9,9-dimethylfluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 320.24
Molecular Formula: C21H25BO2
InChi Key: DAZFRJAIIUPRQZ-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC2=C(C=C1)C1=CC=CC=C1C2(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results