accessibility menu, dialog, popup

UserName

CAS RN 5698-98-6

MgOOH2COOH2C

CAS RN 5698-98-6

IUPAC Name: magnesium(2+) bis(prop-2-enoate)
Synonyms: magnesium(2+) diacrylate
Molecular Weight (g/mol): 166.42
Molecular Formula: C6H6MgO4
InChi Key: DWLAVVBOGOXHNH-UHFFFAOYSA-L
SMILES: [Mg++].[O-]C(=O)C=C.[O-]C(=O)C=C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)