accessibility menu, dialog, popup

UserName

NHNN

CAS RN 57090-88-7

IUPAC Name: 1H-imidazole-5-carbonitrile
Synonyms: 3H-imidazole-4-carbonitrile
Molecular Weight (g/mol): 93.09
Molecular Formula: C4H3N3
InChi Key: NWVGXXPWOYZODV-UHFFFAOYSA-N
SMILES: N#CC1=CN=CN1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results