accessibility menu, dialog, popup

UserName

CH3H3CCH3OONNNNOO

CAS RN 571189-16-7

IUPAC Name: tert-butyl 4-(6-nitropyridin-3-yl)piperazine-1-carboxylate
Synonyms: tert-butyl 4-(6-nitropyridin-3-yl)piperazine-1-carboxylate
Molecular Weight (g/mol): 308.34
Molecular Formula: C14H20N4O4
InChi Key: SUWKOEMQNOBJEQ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCN(CC1)C1=CN=C(C=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results