accessibility menu, dialog, popup

UserName

CAS RN 573-03-5

OHOF

CAS RN 573-03-5

IUPAC Name: 4-fluoronaphthalene-1-carboxylic acid
Synonyms: 4-fluoronaphthalene-1-carboxylic acid
Molecular Weight (g/mol): 190.17
Molecular Formula: C11H7FO2
InChi Key: DEWIOKQDRWFLFW-UHFFFAOYSA-N
SMILES: OC(=O)C1=C2C=CC=CC2=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results