CAS RN 573-20-6
CAS RN 573-20-6
IUPAC Name:
4-(acetyloxy)-2-methylnaphthalen-1-yl acetate
Synonyms:
vitamin K4 adaprin kapathrom kapilin kapilon vitamin K diacetate kappaxan davitamon-K kativ powder davitamon-K-oral
Molecular Weight (g/mol):
258.27
Molecular Formula:
C15H14O4
InChi Key:
RYWSYCQQUDFMAU-UHFFFAOYSA-N
SMILES:
CC(=O)OC1=C2C=CC=CC2=C(OC(C)=O)C(C)=C1
1
–
3
of
3
results
1
Acetomenaphthone (Menadiol Diacetate), LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
2
Acetomenaphthone (Menadiol Diacetate), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
Vitamin K4, MedChemExpress
MedChemExpress Vitamin K4 is a chemically synthesized Vitamin K which plays an important role in the normal blood coagulation system.