accessibility menu, dialog, popup

UserName

OOF

CAS RN 5731-10-2

IUPAC Name: 4'-fluoro-[1,1'-biphenyl]-4-carboxylate
Synonyms: 4'-fluoro-[1,1'-biphenyl]-4-carboxylate
Molecular Weight (g/mol): 215.2
Molecular Formula: C13H8FO2
InChi Key: LXWNTLBMNCXRQN-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=C(C=C1)C1=CC=C(F)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)