CAS RN 575-04-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
7-Methoxy-4-(trifluoromethyl)coumarin, TRC
CAS: 575-04-2 Molecular Formula: C8 H8 Br N3 Molecular Weight (g/mol): 226.073 Synonym: 7-Methoxy-4-(trifluoromethyl)-2H-1-benzopyran-2-one,4-(Trifluoromethyl)-7-methoxycoumarin,7-Methoxy-4-trifluoromethylcoumarin IUPAC Name: 5-bromo-1-methylindazol-3-amine SMILES: Cn1nc(N)c2cc(Br)ccc12