CAS RN 57677-80-2
IUPAC Name:
2-(4-methoxyphenyl)-1,3-thiazole-4-carboxylate
Synonyms:
2-(4-methoxyphenyl)-1,3-thiazole-4-carboxylate
Molecular Weight (g/mol):
234.25
Molecular Formula:
C11H8NO3S
InChi Key:
OIBLFOXWCGIRIQ-UHFFFAOYSA-M
SMILES:
COC1=CC=C(C=C1)C1=NC(=CS1)C([O-])=O
1
–
1
of
1
results
1
2-(4-Methoxyphenyl)thiazole-4-carboxylic acid, 97%, Thermo Scientific™
CAS: 57677-80-2 Molecular Formula: C11H9NO3S Molecular Weight (g/mol): 235.257 MDL Number: MFCD03791177 InChI Key: OIBLFOXWCGIRIQ-UHFFFAOYSA-N Synonym: 2-4-methoxyphenyl thiazole-4-carboxylic acid,2-4-methoxyphenyl-1,3-thiazole-4-carboxylic acid,2-4-methoxy-phenyl-thiazole-4-carboxylic acid,2-4-methoxyphenyl-4-thiazolecarboxylic acid,acmc-209lzq,2-4-methoxyphenyl thiazole-4-carboxylicacid,4-thiazolecarboxylicacid, 2-4-methoxyphenyl,4-thiazolecarboxylic acid, 2-4-methoxyphenyl PubChem CID: 2763565 IUPAC Name: 2-(4-methoxyphenyl)-1,3-thiazole-4-carboxylic acid SMILES: COC1=CC=C(C=C1)C2=NC(=CS2)C(=O)O