CAS RN 5769-15-3
CAS RN 5769-15-3
IUPAC Name:
4-chlorobenzene-1-sulfonyl isocyanate
Synonyms:
4-chlorobenzenesulfonyl isocyanate
Molecular Weight (g/mol):
217.62
Molecular Formula:
C7H4ClNO3S
InChi Key:
JGHDVROWMPBQSR-UHFFFAOYSA-N
SMILES:
ClC1=CC=C(C=C1)S(=O)(=O)N=C=O
1
–
2
of
2
results
1
4-Chlorobenzenesulfonyl Isocyanate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-Chlorobenzenesulfonyl Isocyanate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.