CAS RN 57817-27-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
8-Propyl Etodolac, TRC
CAS: 57817-27-3 Molecular Formula: C18H23NO3 Molecular Weight (g/mol): 301.38 Synonym: 1-Ethyl-1,3,4,9-tetrahydro-8-propyl-pyrano[3,4-b]indole-1-acetic Acid,NSC 282127,Etodolac Impurity E (EP),Etodolac EP Impurity E IUPAC Name: 2-(1-ethyl-8-propyl-4,9-dihydro-3H-pyrano[3,4-b]indol-1-yl)acetic acid SMILES: CCCc1cccc2c3CCOC(CC)(CC(=O)O)c3[nH]c12