accessibility menu, dialog, popup

UserName

CAS RN 5788-58-9

IUPAC Name: 4,5-dibromo-2,3-dihydropyridazin-3-one
Synonyms: 4,5-dibromo-2H-pyridazin-3-one
Molecular Weight (g/mol): 253.88
Molecular Formula: C4H2Br2N2O
InChi Key: AGLQURQNVJVJNB-UHFFFAOYSA-N
SMILES: BrC1=C(Br)C(=O)NN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results