accessibility menu, dialog, popup

UserName

CAS RN 579476-26-9

OHONN

CAS RN 579476-26-9

IUPAC Name: 3-(pyrimidin-2-yl)benzoic acid
Synonyms: 3-(pyrimidin-2-yl)benzoic acid
Molecular Weight (g/mol): 200.2
Molecular Formula: C11H8N2O2
InChi Key: KLTNGIZIUGYITR-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(=C1)C1=NC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results