accessibility menu, dialog, popup

UserName

CAS RN 58-98-0

OHOHOOPHOOOPHOHOONOHNO

CAS RN 58-98-0

IUPAC Name: [({[5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl)oxy]phosphonic acid
Synonyms: uridine diphosphate
Molecular Weight (g/mol): 404.16
Molecular Formula: C9H14N2O12P2
InChi Key: XCCTYIAWTASOJW-UHFFFAOYNA-N
SMILES: OC1C(O)C(OC1COP(O)(=O)OP(O)(O)=O)N1C=CC(=O)NC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results