accessibility menu, dialog, popup

UserName

CAS RN 5811-88-1

ONH2OOH

CAS RN 5811-88-1

IUPAC Name: 8-carbamoylnaphthalene-1-carboxylic acid
Synonyms: 8-carbamoylnaphthalene-1-carboxylic acid
Molecular Weight (g/mol): 215.21
Molecular Formula: C12H9NO3
InChi Key: GRVIMBYDMARKEZ-UHFFFAOYSA-N
SMILES: NC(=O)C1=C2C(C=CC=C2C(O)=O)=CC=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results