accessibility menu, dialog, popup

UserName

H3CCH3

CAS RN 582-16-1

IUPAC Name: 2,7-dimethylnaphthalene
Synonyms: 2,7-dimethylnaphthalene
Molecular Weight (g/mol): 156.23
Molecular Formula: C12H12
InChi Key: LRQYSMQNJLZKPS-UHFFFAOYSA-N
SMILES: CC1=CC2=CC(C)=CC=C2C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results