CAS RN 5837-78-5
CAS RN 5837-78-5
IUPAC Name:
ethyl (2E)-2-methylbut-2-enoate
Synonyms:
ethyl tiglate tiglic acid, ethyl ester (6CI,7CI) ethyl ester (6CI,7CI) tiglic acid
Molecular Weight (g/mol):
128.17
Molecular Formula:
C7H12O2
InChi Key:
OAPHLAAOJMTMLY-GQCTYLIASA-N
SMILES:
CCOC(=O)C(\C)=C\C
1
–
1
of
1
results
1
Ethyl tiglate, 98%
CAS: 5837-78-5 Molecular Formula: C7H12O2 Molecular Weight (g/mol): 128.171 MDL Number: MFCD00015183 InChI Key: OAPHLAAOJMTMLY-GQCTYLIASA-N Synonym: ethyl tiglate,tiglic acid, ethyl ester,ethyl 2-methylcrotonate,ethyl alpha-methylcrotonate,ethyl trans-2-methyl-2-butenoate,e-2-methyl-2-butenoic acid ethyl ester,ethyl e-2-methylcrotonate,ethyl trans-2-methylcrotonate,tiglic acid ethyl ester,ethyl 2-methylbut-2-enoate PubChem CID: 5281163 ChEBI: CHEBI:4892 IUPAC Name: ethyl (E)-2-methylbut-2-enoate SMILES: CCOC(=O)C(=CC)C