CAS RN 5838-34-6
CAS RN 5838-34-6
IUPAC Name:
4-[2-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)acetamido]-N-(1,3-thiazol-2-yl)benzamide
Synonyms:
4-[2-(1,3-dioxoisoindol-2-yl)acetamido]-N-(1,3-thiazol-2-yl)benzamide
Molecular Weight (g/mol):
406.42
Molecular Formula:
C20H14N4O4S
InChi Key:
RFNALTSHRWGOHV-UHFFFAOYSA-N
SMILES:
O=C(CN1C(=O)C2=CC=CC=C2C1=O)NC1=CC=C(C=C1)C(=O)NC1=NC=CS1
1
–
1
of
1
results
1
Sodium 1-undecanesulfonate, 98+% (dry wt.), water <2%
CAS: 5838-34-6 Molecular Formula: C20H14N4O4S Molecular Weight (g/mol): 406.42 MDL Number: MFCD00053627 InChI Key: RFNALTSHRWGOHV-UHFFFAOYSA-N Synonym: sodium 1-undecanesulfonate,sodium undecane-1-sulfonate,1-undecanesulfonic acid sodium salt,sodium undecylsulfonate,ipc-alks-11,acmc-1bn99,1-undecanesulfonicacid, sodium salt 1:1 PubChem CID: 23692564 IUPAC Name: sodium;undecane-1-sulfonate SMILES: O=C(CN1C(=O)C2=CC=CC=C2C1=O)NC1=CC=C(C=C1)C(=O)NC1=NC=CS1