accessibility menu, dialog, popup

UserName

CAS RN 585-36-4

IUPAC Name: 3-(trifluoromethyl)cyclohexan-1-one
Synonyms: 3-(trifluoromethyl)cyclohexan-1-one
Molecular Weight (g/mol): 166.14
Molecular Formula: C7H9F3O
InChi Key: YHADNJOEQWBXAR-UHFFFAOYNA-N
SMILES: FC(F)(F)C1CCCC(=O)C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results