accessibility menu, dialog, popup

UserName

CAS RN 586-61-8

IUPAC Name: 1-bromo-4-(propan-2-yl)benzene
Synonyms: 1-isopropyl-4-bromobenzene
Molecular Weight (g/mol): 199.09
Molecular Formula: C9H11Br
InChi Key: MOZHUOIQYVYEPN-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C(Br)C=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)