CAS RN 58648-57-0
CAS RN 58648-57-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Cefamandole Lithium, TRC
CAS: 58648-57-0 Molecular Formula: C18 H17 N6 O5 S2 . Li Molecular Weight (g/mol): 468.44 Synonym: 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[(hydroxyphenylacetyl)amino]-3-[[(1-methyl-1H-tetrazol-5-yl)thio]methyl]-8-oxo-, monolithium salt, [6R-[6α,7β(R*)]]- (9CI),Cefamandole lithium,Cefamandole lithium salt IUPAC Name: lithium;(6R,7R)-7-[[(2R)-2-hydroxy-2-phenylacetyl]amino]-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate SMILES: [Li+].Cn1nnnc1SCC2=C(N3[C@H](SC2)[C@H](NC(=O)[C@H](O)c4ccccc4)C3=O)C(=O)[O-]