accessibility menu, dialog, popup

UserName

CAS RN 587-02-0

H3CH2N

CAS RN 587-02-0

IUPAC Name: 3-ethylaniline
Synonyms: m-ethylaniline
Molecular Weight (g/mol): 121.18
Molecular Formula: C8H11N
InChi Key: AMKPQMFZCBTTAT-UHFFFAOYSA-N
SMILES: CCC1=CC=CC(N)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results