CAS RN 58775-05-6
IUPAC Name:
2,7-di-tert-butyl-9H-fluorene
Synonyms:
2,7-di-tert-butyl-9H-fluorene
Molecular Weight (g/mol):
278.44
Molecular Formula:
C21H26
InChi Key:
DFZYPLLGAQIQTD-UHFFFAOYSA-N
SMILES:
CC(C)(C)C1=CC2=C(C=C1)C1=C(C2)C=C(C=C1)C(C)(C)C
1
–
1
of
1
results
1
2,7-Di-tert-butylfluorene, 97+%
CAS: 58775-05-6 Molecular Formula: C21H26 Molecular Weight (g/mol): 278.439 MDL Number: MFCD03093998 InChI Key: DFZYPLLGAQIQTD-UHFFFAOYSA-N Synonym: 2,7-di-tert-butylfluorene,2,7-di-tert-butyl-9h-fluorene,2,7-di-t-butylfluorene,9h-fluorene, 2,7-bis 1,1-dimethylethyl,pubchem15066,2,7-di-tert-butyl fluorene,2,7-di-tert-butyl-fluorene,ksc491i2b PubChem CID: 4090082 IUPAC Name: 2,7-ditert-butyl-9H-fluorene SMILES: CC(C)(C)C1=CC2=C(C=C1)C3=C(C2)C=C(C=C3)C(C)(C)C