accessibility menu, dialog, popup

UserName

HOOOClCl

CAS RN 588-22-7

IUPAC Name: 2-(3,4-dichlorophenoxy)acetic acid
Synonyms: 3,4-dichlorophenoxyacetic acid
Molecular Weight (g/mol): 221.03
Molecular Formula: C8H6Cl2O3
InChi Key: SNYRXHULAWEECU-UHFFFAOYSA-N
SMILES: OC(=O)COC1=CC=C(Cl)C(Cl)=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results