CAS RN 59-85-8
CAS RN 59-85-8
IUPAC Name:
(4-carboxyphenyl)mercurylium chloride
Synonyms:
4-chloromercuribenzoic acid
Molecular Weight (g/mol):
357.16
Molecular Formula:
C7H5ClHgO2
InChi Key:
YFZOUMNUDGGHIW-UHFFFAOYSA-M
SMILES:
[Cl-].OC(=O)C1=CC=C([Hg+])C=C1
1
–
1
of
1
results
1
4-Chloromercuribenzoic Acid, TRC
CAS: 59-85-8 Molecular Formula: C7 H5 Cl Hg O2 Molecular Weight (g/mol): 357.16 IUPAC Name: (4-carboxyphenyl)-chloromercury SMILES: OC(=O)c1ccc([Hg]Cl)cc1