accessibility menu, dialog, popup

UserName

CAS RN 591-18-4

BrI

CAS RN 591-18-4

IUPAC Name: 1-bromo-3-iodobenzene
Synonyms: benzene, 1-bromo-3-iodo-
Molecular Weight (g/mol): 282.91
Molecular Formula: C6H4BrI
InChi Key: CTPUUDQIXKUAMO-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(I)=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)