accessibility menu, dialog, popup

UserName

CAS RN 593-79-3

H3CSeCH3

CAS RN 593-79-3

IUPAC Name: (methylselanyl)methane
Synonyms: dimethylselenide dimethylselenide, 75se-labeled dimethyl-selenide dme-selenide 75se-labeled dimethylselenide C2H6se selenide, dimethyl-
Molecular Weight (g/mol): 109.04
Molecular Formula: C2H6Se
InChi Key: RVIXKDRPFPUUOO-UHFFFAOYSA-N
SMILES: C[Se]C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)