accessibility menu, dialog, popup

UserName

H3CSOOO

CAS RN 594-45-6

IUPAC Name: ethanesulfonate
Synonyms: ethanesulfonate ethanesulfonic acid anion ethanesulfonate anion aethansulfonat
Molecular Weight (g/mol): 109.12
Molecular Formula: C2H5O3S
InChi Key: CCIVGXIOQKPBKL-UHFFFAOYSA-M
SMILES: CCS([O-])(=O)=O
Molecular Weight (g/mol) 
  • (8)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (5)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (3)
  • (8)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (5)
  • (3)
  • (3)
  • (3)
  • (2)
  • (6)