CAS RN 594-56-9
IUPAC Name:
2,3,3-trimethylbut-1-ene
Synonyms:
2,3,3-trimethyl-1-butene
Molecular Weight (g/mol):
98.19
Molecular Formula:
C7H14
InChi Key:
AUYRUAVCWOAHQN-UHFFFAOYSA-N
SMILES:
CC(=C)C(C)(C)C
1
–
1
of
1
results
1
2,3,3-Trimethyl-1-butene, 99+%
CAS: 594-56-9 Molecular Formula: C7H14 Molecular Weight (g/mol): 98.19 MDL Number: MFCD00008850 InChI Key: AUYRUAVCWOAHQN-UHFFFAOYSA-N Synonym: 2,3,3-trimethyl-1-butene,1-butene, 2,3,3-trimethyl,triptene,acmc-209mdj,2,3-trimethyl-1-butene,1-butene,3,3-trimethyl,1-butene,2,3,3-trimethyl,ch3 3cc ch3 =ch2 PubChem CID: 11669 IUPAC Name: 2,3,3-trimethylbut-1-ene SMILES: CC(=C)C(C)(C)C