accessibility menu, dialog, popup

UserName

CAS RN 5941-07-1

H2ONaOHHNOSOOOSHOOO(E)

CAS RN 5941-07-1

IUPAC Name: sodium 5-hydroxy-4-({[(1E)-6-oxocyclohexa-2,4-dien-1-ylidene]methyl}amino)-7-sulfonaphthalene-2-sulfonate hydrate
Synonyms: sodium 5-hydroxy-4-({[(1E)-6-oxocyclohexa-2,4-dien-1-ylidene]methyl}amino)-7-sulfonaphthalene-2-sulfonate hydrate
Molecular Weight (g/mol): 463.41
Molecular Formula: C17H14NNaO9S2
InChi Key: BEDIAJJRAYEWBI-TTWKNDKESA-M
SMILES: O.[Na+].OC1=C2C(N\C=C3/C=CC=CC3=O)=CC(=CC2=CC(=C1)S(O)(=O)=O)S([O-])(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results