accessibility menu, dialog, popup

UserName

CH3H3COH

CAS RN 599-64-4

IUPAC Name: 4-(2-phenylpropan-2-yl)phenol
Synonyms: p-cumylphenol 4-(1-methyl-1-phenethyl)phenol p-(α-cumenyl)phenol
Molecular Weight (g/mol): 212.29
Molecular Formula: C15H16O
InChi Key: QBDSZLJBMIMQRS-UHFFFAOYSA-N
SMILES: CC(C)(C1=CC=CC=C1)C1=CC=C(O)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)