accessibility menu, dialog, popup

UserName

CAS RN 6001-74-7

CH3SNClHCl

CAS RN 6001-74-7

IUPAC Name: 5-(2-chloroethyl)-4-methyl-1,3-thiazole hydrochloride
Synonyms: chlormethiazole hydrochloride
Molecular Weight (g/mol): 198.11
Molecular Formula: C6H9Cl2NS
InChi Key: OFXYKSLKNMTBHK-UHFFFAOYSA-N
SMILES: Cl.CC1=C(CCCl)SC=N1
Quantity 
  • (1)
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results