accessibility menu, dialog, popup

UserName

NH2OHNOO

CAS RN 603-85-0

IUPAC Name: 2-amino-3-nitrophenol
Synonyms: phenol, 2-amino-3-nitro-
Molecular Weight (g/mol): 154.13
Molecular Formula: C6H6N2O3
InChi Key: KUCWUAFNGCMZDB-UHFFFAOYSA-N
SMILES: NC1=C(O)C=CC=C1[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)