accessibility menu, dialog, popup

UserName

H3CCH3CH3H3CH3COO

CAS RN 604-32-0

IUPAC Name: 9a,11a-dimethyl-1-(6-methylheptan-2-yl)-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-yl benzoate
Synonyms: cholesteryl benzoate
Molecular Weight (g/mol): 490.77
Molecular Formula: C34H50O2
InChi Key: UVZUFUGNHDDLRQ-UHFFFAOYNA-N
SMILES: CC(C)CCCC(C)C1CCC2C3CC=C4CC(CCC4(C)C3CCC12C)OC(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results