accessibility menu, dialog, popup

UserName

H3CNHH3CNHCH3CH3

CAS RN 60558-96-5

IUPAC Name: [3-(diethylazaniumyl)propyl]diethylazanium
Synonyms: [3-(diethylammonio)propyl]diethylazanium
Molecular Weight (g/mol): 188.36
Molecular Formula: C11H28N2
InChi Key: RCZLVPFECJNLMZ-UHFFFAOYSA-P
SMILES: CC[NH+](CC)CCC[NH+](CC)CC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results