accessibility menu, dialog, popup

UserName

CAS RN 607378-18-7

OHNNHNNHOHHClHCl

CAS RN 607378-18-7

IUPAC Name: 2-(adamantan-1-yl)-N-[2-({2-[(2-hydroxyethyl)amino]ethyl}amino)quinolin-5-yl]acetamide dihydrochloride
Synonyms: 2-(adamantan-1-yl)-N-[2-({2-[(2-hydroxyethyl)amino]ethyl}amino)quinolin-5-yl]acetamide dihydrochloride
Molecular Weight (g/mol): 495.49
Molecular Formula: C25H36Cl2N4O2
InChi Key: BVFONFUUWORSPO-UHFFFAOYSA-N
SMILES: Cl.Cl.OCCNCCNC1=NC2=CC=CC(NC(=O)CC34CC5CC(CC(C5)C3)C4)=C2C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results