accessibility menu, dialog, popup

UserName

NH2ClCl

CAS RN 608-27-5

IUPAC Name: 2,3-dichloroaniline
Synonyms: 2,3-dichloroaniline
Molecular Weight (g/mol): 162.01
Molecular Formula: C6H5Cl2N
InChi Key: BRPSAOUFIJSKOT-UHFFFAOYSA-N
SMILES: NC1=CC=CC(Cl)=C1Cl
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)