accessibility menu, dialog, popup

UserName

HCNOO

CAS RN 6097-08-1

IUPAC Name: 2-(hex-5-yn-1-yl)-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 2-(hex-5-yn-1-yl)isoindole-1,3-dione
Molecular Weight (g/mol): 227.26
Molecular Formula: C14H13NO2
InChi Key: WALVSJIWVWDFCU-UHFFFAOYSA-N
SMILES: O=C1N(CCCCC#C)C(=O)C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results