accessibility menu, dialog, popup

UserName

CAS RN 6104-59-2

NaCH3ONHCNH3CSOOONCH3SOOO

CAS RN 6104-59-2

IUPAC Name: sodium {4-[(4-ethoxyphenyl)amino]phenyl}bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)methylium
Synonyms: coomassie brilliant blue
Molecular Weight (g/mol): 825.97
Molecular Formula: C45H44N3NaO7S2
InChi Key: HAEGGLCXLFWTBE-UHFFFAOYSA-M
SMILES: [Na+].CCOC1=CC=C(NC2=CC=C(C=C2)[C+](C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C=C1
Color 
  • (2)
  • (1)
  • (4)
  • (4)
  • (2)
  • (1)
Quantity 
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
Grade 
  • (4)
pH 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (4)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (4)
  • (4)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (1)