accessibility menu, dialog, popup

UserName

CAS RN 611-17-6

ClBr

CAS RN 611-17-6

IUPAC Name: 1-(bromomethyl)-2-chlorobenzene
Synonyms: 1-(bromomethyl)-2-chlorobenzene
Molecular Weight (g/mol): 205.48
Molecular Formula: C7H6BrCl
InChi Key: PURSZYWBIQIANP-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=C1CBr
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)