accessibility menu, dialog, popup

UserName

CH3N

CAS RN 611-64-3

IUPAC Name: 9-methylacridine
Synonyms: acridine, 9-methyl-
Molecular Weight (g/mol): 193.25
Molecular Formula: C14H11N
InChi Key: FLDRLXJNISEWNZ-UHFFFAOYSA-N
SMILES: CC1=C2C=CC=CC2=NC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (5)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)