accessibility menu, dialog, popup

UserName

CAS RN 612-14-6

HOOH

CAS RN 612-14-6

IUPAC Name: [2-(hydroxymethyl)phenyl]methanol
Synonyms: 1,2-benzenedimethanol
Molecular Weight (g/mol): 138.17
Molecular Formula: C8H10O2
InChi Key: XMUZQOKACOLCSS-UHFFFAOYSA-N
SMILES: OCC1=CC=CC=C1CO
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)