accessibility menu, dialog, popup

UserName

CAS RN 614-39-1

HClH3CNCH3NHONH2

CAS RN 614-39-1

IUPAC Name: hydrogen 4-amino-N-[2-(diethylamino)ethyl]benzamide chloride
Synonyms: pronestyl procapan procamide procanbid procan procainamide hydrochloride
Molecular Weight (g/mol): 271.79
Molecular Formula: C13H22ClN3O
InChi Key: ABTXGJFUQRCPNH-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CCN(CC)CCNC(=O)C1=CC=C(N)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (3)
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results