accessibility menu, dialog, popup

UserName

O(E)(E)

CAS RN 614-57-3

IUPAC Name: (2E,4E)-1,5-diphenylpenta-2,4-dien-1-one
Synonyms: (2E,4E)-1,5-diphenylpenta-2,4-dien-1-one
Molecular Weight (g/mol): 234.3
Molecular Formula: C17H14O
InChi Key: QONKLJMPKWQQFG-HPIZBCMHSA-N
SMILES: O=C(\C=C\C=C\C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results