accessibility menu, dialog, popup

UserName

CAS RN 61478-28-2

CH3H3CCH3OON(S)(S)PP

CAS RN 61478-28-2

IUPAC Name: tert-butyl 4-(diphenylphosphanyl)-2-[(diphenylphosphanyl)methyl]pyrrolidine-1-carboxylate
Synonyms: tert-butyl 4-(diphenylphosphanyl)-2-[(diphenylphosphanyl)methyl]pyrrolidine-1-carboxylate
Molecular Weight (g/mol): 553.62
Molecular Formula: C34H37NO2P2
InChi Key: BFMKBYZEJOQYIM-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CC(CC1CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results